CAS RN 245124-69-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pipendoxifene hydrochloride, MedChemExpress
MedChemExpress Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) that can be used for the research of breast cancer.