CAS RN 2448414-48-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BRD7-IN-1, MedChemExpress
MedChemExpress BRD7-IN-1, a modified derivative of BI7273 (BRD7/9 inhibitor), binds to a VHL ligand via a linker to form a PROTAC VZ185 (VZ185 against BRD7/9 with DC50s of 4.5 and 1.8 nM, respectively).