CAS RN 2444764-03-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GKI-1, MedChemExpress
MedChemExpress GKI-1 is a Greatwall (GWL) kinase inhibitor with IC50s of 4.9 and 2.5 μM against hGWLFL and hGWL-KinDom, respectively. GKI-1 robustly inhibits ROCK1 with an IC50 of 11 μM, but only weakly affected PKA.