CAS RN 2443966-86-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Macropa-NH2 hydrochloride, MedChemExpress
MedChemExpress Macropa-NH2 hydrochloride is the precursor of Macropa-NCS. Macropa-NCS is conjugated to Anti-Human HER2 (HY-P9907) as well as to the prostate-specific membrane antigen-targeting compound RPS-070 and is a promising therapeutic radionuclide applied in the treatment of soft-tissue metastases.