CAS RN 24144-92-1
IUPAC Nombre:
1-[2-(3,5-dimethoxyphenyl)ethenyl]-2,4-dimethoxybenzene
Sinónimos:
1-[2-(3,5-dimethoxyphenyl)ethenyl]-2,4-dimethoxybenzene
Peso molecular (g/mol):
300.35
Fórmula molecular:
C18H20O4
InChi Key:
JDBCWSHYEQUBLW-UHFFFAOYSA-N
SMILES:
COC1=CC(OC)=C(C=CC2=CC(OC)=CC(OC)=C2)C=C1
1
–
1
de
1
resultados
1
TMS, MedChemExpress
MedChemExpress TMS ((E)-2,3',4,5'-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. TMS shows a lesser extent inhibitory effect on CYP1A1 (IC5050 =3.1 μM). TMS is a methylated derivative of resveratrol and has anti-cancer activity.