CAS RN 23815-87-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Bradykinin (1-7)
MedChemExpress Bradykinin (1-7) is an amino-truncated Bradykinin peptide. Bradykinin (1-7) is a metabolite of Bradykinin, cleaved by endopeptidase.
2
MedChemExpress Bradykinin (1-7)
MedChemExpress Bradykinin (1-7) is an amino-truncated Bradykinin peptide. Bradykinin (1-7) is a metabolite of Bradykinin, cleaved by endopeptidase.