CAS RN 2365033-54-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ FNIR-Tag, NHS
Near-infrared fluorescent dye for labeling of amines; supplied as NHS ester