CAS RN 2361659-62-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK8612, MedChemExpress
MedChemExpress GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1.