CAS RN 2342583-66-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NCC007, MedChemExpress
MedChemExpress NCC007 is a dual casein kinase Iα (CKIα) and δ (CKIδ) inhibitor with IC50s of 1.8 and 3.6 μM, respectively. NCC007 can be used in research of modulating mammalian circadian rhythms.