CAS RN 233254-24-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tomeglovir, MedChemExpress
MedChemExpress Tomeglovir is a potent anti-CMV agent, inhibiting processing of viral DNA-concatemers, with IC50s of 0.34 μM and 0.039 μM for HCMV and MCMV.