CAS RN 232931-57-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SJG-136, MedChemExpress
MedChemExpress SJG-136 is a DNA cross-linking agent, with an XL50 of 45 nM for pBR322 DNA. SJG-136 has potent antitumor activity.