CAS RN 2315-97-1
IUPAC Nombre:
10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium dinitrate
Sinónimos:
lucigenin
Peso molecular (g/mol):
510.51
Fórmula molecular:
C28H22N4O6
InChi Key:
KNJDBYZZKAZQNG-UHFFFAOYSA-N
SMILES:
[O-][N+]([O-])=O.[O-][N+]([O-])=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12
1
–
1
de
1
resultados
1
MedChemExpress Lucigenin
MedChemExpress Lucigenin(L-6868; NSC-151912) is a chemiluminescent probe used to indicate the presence of endogenously generated superoxide anion radicals in cells.