CAS RN 2307668-79-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
2S,4R-Sacubitril, MedChemExpress
MedChemExpress 2S,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.