CAS RN 229961-45-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AGN 194310, MedChemExpress
MedChemExpress AGN 194310 (VTP-194310) is a high affinity, potent and selective retinioic acid receptors (RARs) pan-antagonist with Kd values of 3 nM, 2 nM, 5 nM for RARα, RARβ, RARγ, respectively.