CAS RN 228579-27-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Promociones disponibles
Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents
Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents
Cyclohexyl maltoside detergents are a class of specialized non-ionic detergents designed for the solubilization, stabilization, and manipulation of biomolecules, particularly membrane proteins.