CAS RN 2255338-25-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SGC-iMLLT, MedChemExpress
MedChemExpress SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively.