CAS RN 2253744-54-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Dooku 1
Reversibly blocks Yoda1 activity; does not affect constitutive Piezo1 activity