CAS RN 2245697-83-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(S,R,S)-AHPC-C4-NH2 hydrochloride, MedChemExpress
MedChemExpress (S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.