CAS RN 2244904-70-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
BAY-293, MedChemExpress
MedChemExpress BAY-293, a valuable chemical probe, blocks RAS activation via disruption of the KRAS-SOS1 interaction with an IC50 of 21 nM. BAY-293 is a potent inhibitor of Son of Sevenless 1 (SOS1). SOS1 is the guanine nucleotide exchange factor (GEF) and activator of RAS.
2
Tocris™ BAY 293
Potent KRas/SOS1 interaction inhibitor