CAS RN 2244684-49-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ CM 11
Homo-PROTAC™ for self-degradation of pVHL30
2
Homo-PROTAC pVHL30 degrader 1, MedChemExpress
MedChemExpress Homo-PROTAC pVHL30 degrader 1 is a potent pVHL30 degrader based on PROTAC, consists of two ligands of von Hippel-Lindau.
3