CAS RN 223666-07-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Collagen proline hydroxylase inhibitor, MedChemExpress
MedChemExpress Collagen proline hydroxylase inhibitor is a collagen proline hydroxylase inhibitor; useful for antifibroproliferative agents.