CAS RN 223652-90-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Garenoxacin Mesylate hydrate, MedChemExpress
MedChemExpress Garenoxacin Mesylate hydrate (BMS284756 Mesylate hydrate) is a novel oral des-fluoro(6) quinolone with potent antimicrobial activity, against common respiratory pathogens, including resistant strains.