CAS RN 223568-55-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EP1013, MedChemExpress
MedChemExpress EP1013 (F1013) is a broad-spectrum caspase selective inhibitor, used in the research of type 1 diabetes.