CAS RN 221877-56-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
42-(2-Tetrazolyl)rapamycin, MedChemExpress
MedChemExpress 42-(2-Tetrazolyl)rapamycin is a prodrug compound of a rapamycin analog extracted from patent US 20080171763 A1, Example 1. Rapamycin is a specific mTOR inhibitor.