CAS RN 2215120-36-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Interferon receptor inducer-1, MedChemExpress
MedChemExpress Interferon receptor inducer-1 (compound 6) is an interferon (IFN) receptor inducer. Used accordingly in the treatment of a disorder in which the induction of interferon is involved.