CAS RN 221174-33-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glyoxalase I inhibitor, MedChemExpress
MedChemExpress Glyoxalase I inhibitor is a potent Glyoxalase I inhibitor, candidate for anticancer agents.