CAS RN 220460-92-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
WAY-151932, MedChemExpress
MedChemExpress WAY-151932 is a vasopressin V2-receptor agonist with IC50 of 80.3 nM and 778 nM in human-V2 binding and V1a binding assay.