CAS RN 2201066-53-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AGI-24512, MedChemExpress
MedChemExpress AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor, with an IC50 of 8 nM. AGI-24512 triggers DNA damage response. AGI-24512 can block proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer.