CAS RN 2201057-10-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MAT2A inhibitor 1, MedChemExpress
MedChemExpress MAT2A inhibitor 1 is a methionine adenosyltransferase 2A (MATA2) inhibitor with an IC50 less than l00 nM.