CAS RN 219832-49-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MP-A08, MedChemExpress
MedChemExpress MP-A08 is a highly selective ATP competitive sphingosine kinase (SPHK1) inhibitor that targets both SphK1 and SphK2 with Ki values of 6.9 ± 0.8 μM and 27 ± 3 μM, respectively.
2
Tocris Bioscience™ MP A08
Selective ATP competitive sphingosine kinase 2 and 1 (SphK2/1) inhibitor
3
Tocris Bioscience™ MP A08
Selective ATP competitive SphK1 and 2 inhibitor; cell permeable