CAS RN 21802-37-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Caerulomycin A, MedChemExpress
MedChemExpress Caerulomycin A (Cerulomycin; Caerulomycin), an antifungal compound, induces generation of T cells, enhances TGF-β-Smad3 protein signaling via suppressing interferon-γ-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases.