CAS RN 2177266-81-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ribocil-C (R enantiomer), MedChemExpress
MedChemExpress Ribocil-C R enantiomer is the R enantiomer of Ribocil-C. Ribocil-C is a highly selective inhibitor of bacterial riboflavin riboswitches.