CAS RN 216974-75-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Bevacizumab
MedChemExpress Bevacizumab, a humanized IgG1 monoclonal antibody, specifically binds to all VEGF-A isoforms with high affinity.
2
MedChemExpress Bevacizumab
MedChemExpress Bevacizumab, a humanized IgG1 monoclonal antibody, specifically binds to all VEGF-A isoforms with high affinity.