CAS RN 215510-22-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Prolactin Releasing Peptide (1-31), human
MedChemExpress Prolactin Releasing Peptide (1-31), human is a high affinity GPR10 ligand that cause the release of the prolactin. Human and rat Prolactin Releasing Peptide (1-31) binds to GPR10 with Kis of 1.03 and 0.33 nM, respectively.
2
MedChemExpress Prolactin Releasing Peptide (1-31), human
MedChemExpress Prolactin Releasing Peptide (1-31), human is a high affinity GPR10 ligand that cause the release of the prolactin. Human and rat Prolactin Releasing Peptide (1-31) binds to GPR10 with Kis of 1.03 and 0.33 nM, respectively.