CAS RN 2140301-96-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Brepocitinib P-Tosylate, MedChemExpress
MedChemExpress Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.