CAS RN 2127150-20-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Janelia Fluor 669, SE
Far red dye (Reactive group: NHS ester). Applications: super resolution microscopy e.g. dSTORM & STED, confocal and live cell imaging
2
Janelia Fluor 669, SE
Far red dye (Reactive group: NHS ester). Applications: super resolution microscopy e.g. dSTORM & STED, confocal and live cell imaging