CAS RN 210829-30-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Gemcitabine elaidate, MedChemExpress
MedChemExpress Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.