CAS RN 210344-98-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Z-IETD-FMK
MedChemExpress Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a selective and cell permeable caspase-8 inhibitor. Z-IETD-FMK is also a granzyme B inhibitor.
2
MedChemExpress Z-IETD-FMK
MedChemExpress Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a selective and cell permeable caspase-8 inhibitor. Z-IETD-FMK is also a granzyme B inhibitor.