CAS RN 2102933-95-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
Tocris Bioscience™ GSK LSD 1 Dihydrochloride
Potent and selective LSD1 inhibitor
2
GSK-LSD1 dihydrochloride, MedChemExpress
MedChemExpress GSK-LSD1 dihydrochloride is a potent, selective and irreversible lysine specific demethylase 1 (LSD1) inhibitor with an IC50 of 16 nM.
3
4