CAS RN 210236-90-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Fluorescein Tyramide
Green fluorescent reagent widely used for signal amplification in IHC and FISH