CAS RN 2098969-71-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DO34 analog, MedChemExpress
MedChemExpress DO34 analog is a triazole DAGL(α) inhibitor extracted from patent WO2017096315 A1, compound 100.