CAS RN 2091134-68-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Golidocitinib, MedChemExpress
MedChemExpress Golidocitinib (AZD4205) is a selective JAK1 inhibitor, with an IC50 of 73 nM, weakly inhibits JAK2 (IC50>14.7 μM), and shows little inhibition on JAK3 (IC50>30 μM).