CAS RN 2070015-29-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SJB3-019A, MedChemExpress
MedChemExpress SJB3-019A is a potent and novel USP1 inhibitor, 5 times more potent than SJB2-043 in promoting ID1 degradation and cytoxicity in K562 cells with IC50 of 0.0781 μM.