CAS RN 2070015-24-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pimelic Diphenylamide 106 (analog), MedChemExpress
MedChemExpress Pimelic Diphenylamide 106 analog is an analog of Pimelic Diphenylamide 106 with unknown biological activity.