CAS RN 2068806-31-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ JBJ-03-142-02
Highly potent EGFR and ErbB2 inhibitor; inhibts wild type and mutated receptors