CAS RN 2068-83-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
3-Hydroxyisobutyric acid, MedChemExpress
MedChemExpress 3-Hydroxyisobutyric acid is an important interorgan metabolite, an intermediate in the pathways of l-valine and thymine and a good gluconeogenic substrate.