CAS RN 2066-89-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pasiniazid, MedChemExpress
MedChemExpress Pasiniazid is an anti-TB and anti-leprosy drug, used to treat various types of TB and leprosy.