CAS RN 2060259-60-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SRT 1720 Hydrochloride, MedChemExpress
MedChemExpress SRT 1720 Hydrochloride is a selective activator of SIRT1 with an EC50 of 0.10 μM, and shows less potent activities on SIRT2 and SIRT3.