CAS RN 20424-00-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Deprodone propionate, MedChemExpress
MedChemExpress Deprodone propionate (RD20000) is a corticosteroid which is obtained by esterifying with propionic acid the 17-position of the prednisolone skeleton and deoxidating its 21-position.