CAS RN 2018300-62-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TPOP146, MedChemExpress
MedChemExpress TPOP146 is a selective CBP/P300 benzoxazepine bromodomain inhibitor with Kd values of 134 nM and 5.02 μM for CBP and BRD4.